Learn More
Cayman Chemical Asoxime (chloride), Purity: ≥95%, CAS: 34433-31-3, 50 mg
SDP
Catalog No. 502979527
Encompass_Preferred
An asymmetric bis-pyridinium aldoxime that functions as an AChE reactivator through nucleophilic attack of oximate anions on organophosphate-AChE conjugates; demonstrates therapeutic activity in experimental models of organophosphate poisoning
Catalog No. 50-297-9527 Supplier Cayman Chemical Supplier No. 1860750 MG
An asymmetric bis-pyridinium aldoxime that functions as an AChE reactivator through nucleophilic attack of oximate anions on organophosphate-AChE conjugates; demonstrates therapeutic activity in experimental models of organophosphate poisoning