Learn More
Medchemexpress LLC ASP-4058 hydrochloride | 952510-14-4 | 99.7% | 478.78 | 25 MG
SDP
Catalog No. 5000345919
Encompass_Preferred
ASP-4058 hydrochloride is a next-generation, selective, and orally active agonist for Sphingosine 1-Phosphate receptors 1 and 5 (S1P1 and S1P5). This compound has been shown to ameliorate rodent experimental autoimmune encephalomyelitis with a favorable safety profile.
  • Next-generation S1P1 and S1P5 agonist
  • Selective and orally active
  • Ameliorates rodent experimental autoimmune encephalomyelitis
  • Favorable safety profile
Catalog No. 50-003-45919 Supplier Medchemexpress LLC Supplier No. 25MGHY111021A
ASP-4058 hydrochloride is a next-generation, selective, and orally active agonist for Sphingosine 1-Phosphate receptors 1 and 5 (S1P1 and S1P5). This compound has been shown to ameliorate rodent experimental autoimmune encephalomyelitis with a favorable safety profile.
  • Next-generation S1P1 and S1P5 agonist
  • Selective and orally active
  • Ameliorates rodent experimental autoimmune encephalomyelitis
  • Favorable safety profile