Learn More
TARGETMOL CHEMICALS INC ASP-9521 25MG
SDP
Catalog No. 502878686
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes.ASP-9521 is a selective, potent and orally active indole-based AKR1C3 inhibitor with an IC50 of 11 nM for human AKR1C3. Purity 96.96%
Catalog No. 50-287-8686 Supplier TARGETMOL CHEMICALS INC Supplier No. T452925MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes.ASP-9521 is a selective, potent and orally active indole-based AKR1C3 inhibitor with an IC50 of 11 nM for human AKR1C3. Purity 96.96%