Learn More
Medchemexpress LLC Asp-AMS | 828288-98-8 | 98.14% | 461.41 | 50 MG
SDP
Catalog No. 5000437578
Encompass_Preferred
Asp-AMS, an analogue of aspartyl-adenylate, is an aspartyl-tRNA synthetase inhibitor and also a strong competitive inhibitor of the mitochondrial enzyme. It is for research use only.
  • An analogue of aspartyl-adenylate
  • Aspartyl-tRNA synthetase inhibitor
  • Strong competitive inhibitor of the mitochondrial enzyme
  • 500-fold stronger competitive inhibitor of the mitochondrial enzyme than aspartol-AMP (10 nM)
  • 35-fold lower competitor of human and bovine cyt-AspRSs (300 nM)
  • Strong inhibitor with Ki in the nanomolar (nM) range
  • Highest inhibitory effect for the mitochondrial enzyme
  • Most active inhibitor with Ki values in the nanomolar range, with a stronger effect on bacterial AspRSs (E. coli and P. aeruginosa) than on human cytosolic AspRS
Catalog No. 50-004-37578 Supplier Medchemexpress LLC Supplier No. HY11286050MG
Asp-AMS, an analogue of aspartyl-adenylate, is an aspartyl-tRNA synthetase inhibitor and also a strong competitive inhibitor of the mitochondrial enzyme. It is for research use only.
  • An analogue of aspartyl-adenylate
  • Aspartyl-tRNA synthetase inhibitor
  • Strong competitive inhibitor of the mitochondrial enzyme
  • 500-fold stronger competitive inhibitor of the mitochondrial enzyme than aspartol-AMP (10 nM)
  • 35-fold lower competitor of human and bovine cyt-AspRSs (300 nM)
  • Strong inhibitor with Ki in the nanomolar (nM) range
  • Highest inhibitory effect for the mitochondrial enzyme
  • Most active inhibitor with Ki values in the nanomolar range, with a stronger effect on bacterial AspRSs (E. coli and P. aeruginosa) than on human cytosolic AspRS