Learn More
Foxx Life Sciences Aspirator Bottle 20L, PP, White Cap with Spigot
GREENER_CHOICE
Catalog No. NC1818190
Encompass_Preferred
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene laboratory Aspirator Bottles comes with strong easy grip handle and Spigot. Our versatile Bottles are leak proof and aim to take full advantage of storage efficiency at minimal effort. Overall functionality is available for many applications, in 5 Liter, 10 Liter or 20 Liter sizes, with spigots and a Polypropylene (PP) Cap. Foxx laboratory Polypropylene (PP) Aspirator Bottles are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).Cap and Spigot should be removed before autoclaving this Aspirator Bottle.
Catalog No. NC1818190 Supplier Foxx Life Sciences Supplier No. 155L897FLS
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene laboratory Aspirator Bottles comes with strong easy grip handle and Spigot. Our versatile Bottles are leak proof and aim to take full advantage of storage efficiency at minimal effort. Overall functionality is available for many applications, in 5 Liter, 10 Liter or 20 Liter sizes, with spigots and a Polypropylene (PP) Cap. Foxx laboratory Polypropylene (PP) Aspirator Bottles are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).Cap and Spigot should be removed before autoclaving this Aspirator Bottle.