Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 12 Channel, 12 Well, Standard, 19 Gauge, SS Tubes, 7mm Long, 9mm Spacing, Polypropylene
Catalog No. NC2085785
Encompass
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 7mm Long, 9mm Center Spacing, Polypropylene
This item is not returnable. View return policy
Catalog No. NC2085785 Supplier V&P Scientific Inc Supplier No. VP 187PP1S
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 7mm Long, 9mm Center Spacing, Polypropylene