Learn More
Medchemexpress LLC HY-110176 10mg Medchemexpress, ASP2535 CAS:374886-51-8 Purity:>98%
SDP
Catalog No. 501968762
Encompass_Preferred
Medchemexpress, HY-110176 10mg ASP2535 CAS:374886-51-8 ASP2535 is a potent, orally bioavailable, selective, brain permeable and centrally-active glycine transporter-1 (GlyT1) inhibitor. ASP2535 can improve cognitive impairment in animal models of schizophrenia and Alzheimer's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8762 Supplier Medchemexpress LLC Supplier No. HY11017610MG
Medchemexpress, HY-110176 10mg ASP2535 CAS:374886-51-8 ASP2535 is a potent, orally bioavailable, selective, brain permeable and centrally-active glycine transporter-1 (GlyT1) inhibitor. ASP2535 can improve cognitive impairment in animal models of schizophrenia and Alzheimer's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.