Learn More
Apexbio Technology LLC ASP3026 1097917-15-1 200mg
SDP
Catalog No. 5000092980
Encompass_Preferred
ASP3026 (CAS 1097917-15-1) is a selective inhibitor of anaplastic lymphoma kinase (ALK) exhibiting an IC50 of 3 5 nM for ALK enzymatic inhibition When evaluated across a panel of 86 tyrosine kinases ASP3026 demonstrated highest selectivity for ALK ROS and ACK kinases In cellular studies ASP3026 inhibited the proliferation of EML4-ALK-expressing NCI-H2228 non-small cell lung cancer (NSCLC) cells (IC50 64 8 nM) and reduced viability in NPM-ALK anaplastic large cell lymphoma (ALCL) lines with IC50 values ranging from 0 4 to 2 5 M In vivo oral administration of ASP3026 in mouse xenograft models suppressed ALK phosphorylation and significantly reduced tumor growth ASP3026 serves as a valuable tool for investigating ALK-driven oncogenesis and for preclinical evaluation of ALK-targeted therapies
Catalog No. 50-000-92980 Supplier Apexbio Technology LLC Supplier No. A3193200
ASP3026 (CAS 1097917-15-1) is a selective inhibitor of anaplastic lymphoma kinase (ALK) exhibiting an IC50 of 3 5 nM for ALK enzymatic inhibition When evaluated across a panel of 86 tyrosine kinases ASP3026 demonstrated highest selectivity for ALK ROS and ACK kinases In cellular studies ASP3026 inhibited the proliferation of EML4-ALK-expressing NCI-H2228 non-small cell lung cancer (NSCLC) cells (IC50 64 8 nM) and reduced viability in NPM-ALK anaplastic large cell lymphoma (ALCL) lines with IC50 values ranging from 0 4 to 2 5 M In vivo oral administration of ASP3026 in mouse xenograft models suppressed ALK phosphorylation and significantly reduced tumor growth ASP3026 serves as a valuable tool for investigating ALK-driven oncogenesis and for preclinical evaluation of ALK-targeted therapies