Learn More
Medchemexpress LLC HY-13326 50mg Medchemexpress, ASP3026 CAS:1097917-15-1 Purity:>98%
SDP
Catalog No. 502081333
Encompass_Preferred
Medchemexpress, HY-13326 50mg ASP3026 CAS:1097917-15-1 ASP3026 is a potent, selective and orally active inhibitor of anaplastic lymphoma kinase (ALK). ASP3026 induces apoptosis of tumor cells. ASP3026 can be used for the research of non-small cell lung cancer (NSCLC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1333 Supplier Medchemexpress LLC Supplier No. HY1332650MG
Medchemexpress, HY-13326 50mg ASP3026 CAS:1097917-15-1 ASP3026 is a potent, selective and orally active inhibitor of anaplastic lymphoma kinase (ALK). ASP3026 induces apoptosis of tumor cells. ASP3026 can be used for the research of non-small cell lung cancer (NSCLC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.