Learn More
Cayman Chemical Asp371-TyrosInase369-377 25mg
SDP
Catalog No. 502942981
Encompass_Preferred
A tyrosinase antigen peptide; binds to MHC class I subtype HLA-A*0201-positive T2 cells (IC50 = 1.2 UG/ml) and induces lysis of HLA-A2.1-positive T2 cells incubated with cytotoxic T lymphocytes; immunization with [Asp371]-tyrosinase (369-377)-pulsed, CD40L-activated autologous dendritic cells delays tumor growth in an AAD-expressing B16-F1 melanoma model using transgenic mice expressing HLA-A*0201 linked to AAD
Catalog No. 50-294-2981 Supplier Cayman Chemical Supplier No. 4015225 MG
A tyrosinase antigen peptide; binds to MHC class I subtype HLA-A*0201-positive T2 cells (IC50 = 1.2 UG/ml) and induces lysis of HLA-A2.1-positive T2 cells incubated with cytotoxic T lymphocytes; immunization with [Asp371]-tyrosinase (369-377)-pulsed, CD40L-activated autologous dendritic cells delays tumor growth in an AAD-expressing B16-F1 melanoma model using transgenic mice expressing HLA-A*0201 linked to AAD