Learn More
Medchemexpress LLC Asp4132 tosylate | 1640294-30-9 | 99.8% | C46H51F3N6O8S2 | 10MG
SDP
Catalog No. 5000260408
Encompass_Preferred
ASP4132 is an orally active, potent AMPK activator (EC50 18 nM) that exhibits antineoplastic activity and induces tumor regression in breast cancer xenograft models. It is provided as a solid tosylate salt with high purity and is suitable for in vitro and in vivo research when prepared in appropriate solvents.
  • Potent AMPK activation with low nanomolar EC50.
  • Demonstrated antitumor efficacy in xenograft models.
  • High chemical purity for reproducible results.
  • Solid tosylate salt form for improved handling.
  • High DMSO solubility for in vitro dosing.
  • Multiple pack sizes available for flexible scaling.
Catalog No. 50-002-60408 Supplier Medchemexpress LLC Supplier No. HY13644710MG
Add to Cart
Add to Cart
ASP4132 is an orally active, potent AMPK activator (EC50 18 nM) that exhibits antineoplastic activity and induces tumor regression in breast cancer xenograft models. It is provided as a solid tosylate salt with high purity and is suitable for in vitro and in vivo research when prepared in appropriate solvents.
  • Potent AMPK activation with low nanomolar EC50.
  • Demonstrated antitumor efficacy in xenograft models.
  • High chemical purity for reproducible results.
  • Solid tosylate salt form for improved handling.
  • High DMSO solubility for in vitro dosing.
  • Multiple pack sizes available for flexible scaling.