Learn More
Selleck Chemical LLC ASP5878
SDP
Catalog No. 5000002918
Encompass_Preferred
ASP5878 is a novel FGFR-selective inhibitor with IC50 values of 0 47 0 60 0 74 and 3 5 nmol/L for recombinant FGFR1 2 3 and 4 respectively
Catalog No. 50-000-02918 Supplier Selleck Chemical LLC Supplier No. S65392MG
ASP5878 is a novel FGFR-selective inhibitor with IC50 values of 0 47 0 60 0 74 and 3 5 nmol/L for recombinant FGFR1 2 3 and 4 respectively