Learn More
Aviva Systems Biology Corp ASPA Antibody - middle region (ARP56073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASPA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ASPA. Alternate names: ACY2, ASPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61707