Learn More
Medchemexpress LLC Asparagusic acid | 2224-02-4 | 96.0% | C4H6O2S2 | 5 MG
SDP
Catalog No. 5000358317
Encompass_Preferred
Asparagusic acid is a sulfur-containing flavor component derived from Asparagus officinalis Linn. It exhibits an anti-parasitic effect and functions as a plant growth inhibitor. Research indicates that this compound is toxic to several plant parasitic nematodes and can inhibit the growth of lettuce and other seedlings.
  • Sulfur-containing flavor component
  • Derived from Asparagus officinalis Linn.
  • Exhibits anti-parasitic effect
  • Acts as a plant growth inhibitor
  • Toxic to plant parasitic nematodes
  • Inhibits growth in lettuce and other seedlings
Catalog No. 50-003-58317 Supplier Medchemexpress LLC Supplier No. HY507305MG
Add to Cart
Add to Cart
Asparagusic acid is a sulfur-containing flavor component derived from Asparagus officinalis Linn. It exhibits an anti-parasitic effect and functions as a plant growth inhibitor. Research indicates that this compound is toxic to several plant parasitic nematodes and can inhibit the growth of lettuce and other seedlings.
  • Sulfur-containing flavor component
  • Derived from Asparagus officinalis Linn.
  • Exhibits anti-parasitic effect
  • Acts as a plant growth inhibitor
  • Toxic to plant parasitic nematodes
  • Inhibits growth in lettuce and other seedlings