Learn More
Medchemexpress LLC Aspartame | 22839-47-0 | 99.9% | 250 MG
SDP
Catalog No. 5000407402
Encompass_Preferred
Aspartame (Standard) is an analytical standard intended for research and analytical applications. This methyl ester of a dipeptide functions as a synthetic nonnutritive sweetener, appearing as a white to off-white solid. It has a molecular weight of 294.30 and a chemical formula of C14H18N2O5.
  • Reference standard for assay purposes
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
  • Analytical standard grade
Catalog No. 50-004-07402 Supplier Medchemexpress LLC Supplier No. 250MGHYB0361R
Aspartame (Standard) is an analytical standard intended for research and analytical applications. This methyl ester of a dipeptide functions as a synthetic nonnutritive sweetener, appearing as a white to off-white solid. It has a molecular weight of 294.30 and a chemical formula of C14H18N2O5.
  • Reference standard for assay purposes
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
  • Analytical standard grade