Learn More
Biomatik Corporation Aspartate Aminotransferase (Ast) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29502
Encompass_Preferred
Aspartate Aminotransferase (Ast) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Aspartate Aminotransferase (AST). The Aspartate Aminotransferase (Ast) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Met1~Gln413. Applications: WB, IHC, ICC, IP. Target Synonyms: Aspartate Aminotransferase (AST). Accession Number: P17174. Gene ID: 2805. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6759