Learn More
Medchemexpress LLC (-)-Aspartic acid | 1783-96-6 | MFCD00063081 | 98.0% | 133.10 | 5 G
SDP
Catalog No. 5000403551
Encompass_Preferred
(-)-Aspartic acid is a pyroptosis inhibitor. It functions as a neurotransmitter and neuromodulator, participating in hormone synthesis and regulation, and playing a role in the development of the nervous system and endocrine system. It is for research use only and not sold to patients.
  • Pyroptosis inhibitor
  • Neurotransmitter and neuromodulator
  • Participates in hormone synthesis and regulation
  • Plays a role in nervous system development
  • Plays a role in the endocrine system
  • Inhibits HGF cell pyroptosis through the TLRs-NLRP3-caspase 1-GSDMD pathway, preventing gingival fibroblast inflammation
  • Stimulates steroid production in mammalian Leydig cell lines by regulating LH receptor (LHR) exposure on the cell membrane
  • Increases the secretion of gonadotropin-releasing hormone (GnRH), prolactin (PRL), luteinizing hormone (LH), and growth hormone (GH) in rats
Catalog No. 50-004-03551 Supplier Medchemexpress LLC Supplier No. HY420685G
(-)-Aspartic acid is a pyroptosis inhibitor. It functions as a neurotransmitter and neuromodulator, participating in hormone synthesis and regulation, and playing a role in the development of the nervous system and endocrine system. It is for research use only and not sold to patients.
  • Pyroptosis inhibitor
  • Neurotransmitter and neuromodulator
  • Participates in hormone synthesis and regulation
  • Plays a role in nervous system development
  • Plays a role in the endocrine system
  • Inhibits HGF cell pyroptosis through the TLRs-NLRP3-caspase 1-GSDMD pathway, preventing gingival fibroblast inflammation
  • Stimulates steroid production in mammalian Leydig cell lines by regulating LH receptor (LHR) exposure on the cell membrane
  • Increases the secretion of gonadotropin-releasing hormone (GnRH), prolactin (PRL), luteinizing hormone (LH), and growth hormone (GH) in rats