Learn More
Medchemexpress LLC HY-107091 10mg Medchemexpress, Aspartyl-alanyl-diketopiperazine CAS:110954-19-3 Purity:>98%
SDP
Catalog No. 501968364
Encompass_Preferred
Medchemexpress, HY-107091 10mg Aspartyl-alanyl-diketopiperazine CAS:110954-19-3 Aspartyl-alanyl-diketopiperazine (DA-DKP) is an immunomodulatory molecule generated by cleavage and cyclization from the N-terminus of human albumin and can modulate the inflammatory immune response through a molecular pathway implicated in T- lymphocyte anergy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8364 Supplier Medchemexpress LLC Supplier No. HY10709110MG
Medchemexpress, HY-107091 10mg Aspartyl-alanyl-diketopiperazine CAS:110954-19-3 Aspartyl-alanyl-diketopiperazine (DA-DKP) is an immunomodulatory molecule generated by cleavage and cyclization from the N-terminus of human albumin and can modulate the inflammatory immune response through a molecular pathway implicated in T- lymphocyte anergy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.