Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aspartyl-alanyl-diketopiperazine | 110954-19-3 | 98.9% | 186.17 g/mol | C7H10N2O4 | 1 ML
SDP

Catalog No. 5000239764
Encompass_Preferred

Aspartyl-alanyl-diketopiperazine (DA-DKP) is a cyclic dipeptide derived from the N-terminus of human albumin with reported immunomodulatory activity. It modulates inflammatory immune responses and is supplied for laboratory research as a solid or as a ready-to-use solution.

  • Purity 98.87%.
  • Molecular weight 186.17 g/mol.
  • Chemical formula C7H10N2O4.
  • CAS number 110954-19-3.
  • Physical form solid, white to off-white.
  • Also available as a 10 mM solution in DMSO, 1 mL.
  • Typical solid package sizes 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-39764 Supplier Medchemexpress LLC Supplier No. HY10709110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aspartyl-alanyl-diketopiperazine (DA-DKP) is a cyclic dipeptide derived from the N-terminus of human albumin with reported immunomodulatory activity. It modulates inflammatory immune responses and is supplied for laboratory research as a solid or as a ready-to-use solution.

  • Purity 98.87%.
  • Molecular weight 186.17 g/mol.
  • Chemical formula C7H10N2O4.
  • CAS number 110954-19-3.
  • Physical form solid, white to off-white.
  • Also available as a 10 mM solution in DMSO, 1 mL.
  • Typical solid package sizes 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.