Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aspartyl-alanyl-diketopiperazine | 110954-19-3 | 98.9% | 186.17 g/mol | C7H10N2O4 | 50 MG
SDP

Catalog No. 5000239822
Encompass_Preferred

Aspartyl-alanyl-diketopiperazine (DA-DKP) is a cyclic dipeptide derived from the N-terminus of human albumin that modulates inflammatory immune responses and T-lymphocyte anergy. Supplied as a white to off-white solid for research use, the compound includes recommended solubility and in vivo formulation protocols.

  • Immunomodulatory cyclic dipeptide that modulates inflammatory responses.
  • High purity: 98.9% (manufacturer specification).
  • Chemical formula C7H10N2O4 and molecular weight 186.17 g/mol.
  • Appearance: white to off-white solid suitable for research applications.
  • Solubility: highly soluble in DMSO (125 mg/mL); includes in vivo formulation protocols.
  • Available in multiple small research sizes for experimental flexibility.

Catalog No. 50-002-39822 Supplier Medchemexpress LLC Supplier No. HY10709150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aspartyl-alanyl-diketopiperazine (DA-DKP) is a cyclic dipeptide derived from the N-terminus of human albumin that modulates inflammatory immune responses and T-lymphocyte anergy. Supplied as a white to off-white solid for research use, the compound includes recommended solubility and in vivo formulation protocols.

  • Immunomodulatory cyclic dipeptide that modulates inflammatory responses.
  • High purity: 98.9% (manufacturer specification).
  • Chemical formula C7H10N2O4 and molecular weight 186.17 g/mol.
  • Appearance: white to off-white solid suitable for research applications.
  • Solubility: highly soluble in DMSO (125 mg/mL); includes in vivo formulation protocols.
  • Available in multiple small research sizes for experimental flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.