Learn More
Cayman Chemical Aspartyl-alanyl-diketopiperazine, Purity: ≥90%, CAS: 110954-19-3, 10 mg
SDP
Catalog No. 502916777
Encompass_Preferred
An immunosuppressive diketopiperazine derivative of an HSA peptide; reduces antigen-induced TNF-α and IFN-γ production in isolated human T cells at 500 µM
Catalog No. 50-291-6777 Supplier Cayman Chemical Supplier No. 3771510 MG
An immunosuppressive diketopiperazine derivative of an HSA peptide; reduces antigen-induced TNF-α and IFN-γ production in isolated human T cells at 500 µM