Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aspartylglucosaminidase (human recombinant) | >95.0% | 50 UG
SDP

Catalog No. 5000294035
Encompass_Preferred

Recombinant human Aspartylglucosaminidase (AGA) expressed in HEK293 cells, supplied lyophilized with a C-terminal His tag and stabilizing excipients. The protein is purified and characterized for use in biochemical assays, enzyme activity studies, and cell biology applications requiring human AGA.

  • Expression system: HEK293 cells.
  • Tag: C-terminal His tag.
  • Sequence range: residues S24-I346.
  • Purity: >95.0% by reducing SDS-PAGE.
  • Storage: lyophilized from PBS with trehalose, mannitol, and 0.01% Tween 80.
  • Recommended reconstitution: prepare at ≥100 μg/mL in ddH2O.

Catalog No. 50-002-94035 Supplier Medchemexpress LLC Supplier No. HYP7673750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Aspartylglucosaminidase (AGA) expressed in HEK293 cells, supplied lyophilized with a C-terminal His tag and stabilizing excipients. The protein is purified and characterized for use in biochemical assays, enzyme activity studies, and cell biology applications requiring human AGA.

  • Expression system: HEK293 cells.
  • Tag: C-terminal His tag.
  • Sequence range: residues S24-I346.
  • Purity: >95.0% by reducing SDS-PAGE.
  • Storage: lyophilized from PBS with trehalose, mannitol, and 0.01% Tween 80.
  • Recommended reconstitution: prepare at ≥100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.