Learn More
ATCC AsPC-1; Pancreatic Cancer; Human (Homo sapiens)
Catalog No. 502383841
Encompass
AsPC-1 is cell line derived from nude mouse xenografts initiated with cells from the ascites of a 62-year-old, White, female patient with cancer of the pancreas. This cell line can be used in cancer research.
Catalog No. 50-238-3841 Supplier ATCC Supplier No. CRL1682
AsPC-1 is cell line derived from nude mouse xenografts initiated with cells from the ascites of a 62-year-old, White, female patient with cancer of the pancreas. This cell line can be used in cancer research.