Learn More
Bio Serv ASPEN BLOCK LG 1 1/8X1 1/8X3IN
SDP
Catalog No. NC4035877
Encompass_Preferred
Certified Contaminant Screened100 percent US harvested Aspen WoodAspen is a hardwood, but is less dense than other hardwoodsProvides the most natural form of gnawing enrichmentautoclavable
Catalog No. NC4035877 Supplier Bio Serv Supplier No. K3641/ 19
Certified Contaminant Screened100 percent US harvested Aspen WoodAspen is a hardwood, but is less dense than other hardwoodsProvides the most natural form of gnawing enrichmentautoclavable