Learn More
Aspen Surgical™ Securline™ Nonsterile Surgical Skin Marker
Catalog No. 22138865
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
22-138-865 Non-sterile
1 options
Catalog No. 22-138-865 Supplier Aspen Surgical™ Supplier No. 100300PDG
Add to Cart
Add to Cart
  • Nonsterile, nonscratching, with rule
  • Gentian violet ink, surgical prep resistant
  • Nontoxic to epidermal layer of skin
Specifications
Product Type Skin Marker
Tip Style Taper
Sterility Non-sterile
Material (Internal) Gentian Violet
Latex Free Yes
For Use With (Application) Intended for surgical marking on intact skin and helps prevent wrong site surgery, by allowing the surgical site to be marked properly
Quantity 100 Pk.
WARNING: Cancer - www.P65Warnings.ca.gov