Learn More
AdipoGen Aspergillimide (5 mg)
SDP
Supplier:  AdipoGen AGCN20145M005
Encompass_Preferred
Chemical. CAS: 195966-93-9. Formula: C20H29N3O3. MW: 359.5. Isolated from Aspergillus japonicus Fg-551. Antibiotic. Anthelmintic agent. Paralytic against Bombyx mori.
Catalog No. 50-205-3527