Learn More
AdipoGen Aspergillin PZ (5 mg)
SDP
Supplier:  AdipoGen AGCN20162M005
Encompass_Preferred
Chemical. CAS: 483305-08-4. Formula: C24H35NO4. MW: 401.6. Isolated from Aspergillus sp. Antibiotic. Inducer of morphological deformation of the conidia of Pyricularia oryzae.
Catalog No. 50-205-3550