Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Aspergillus brasiliensis derived from ATCC 16404
SDP

Catalog No. 10810324 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-810-324 10 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-810-324 Supplier Microbiologics™ Supplier No. 0392FPC
This item is not returnable. View return policy

EZ-FPC lyophilized QC microorganism preparations are daily process controls for both qualitative and quantitative detection methods. They’re ideal for Statistical Process Charting programs to standardize quality controls throughout your laboratory.

  • Biosafety Level: 1
  • 10 pellets of a quantitative microorganism manufactured as a food process control.fungus; Quantitative Preparations at a concentration of 1.0-9.9E+03 CFU per pellet

Specifications

ATCC® ATCC 16404
Biosafety Level 1
Certifications/Compliance ISO 11133
Content And Storage 2°C to 8°C
Format EZ-FPC
Manufacturing Origin United States
Product Type EZ-FPC
For Use With (Application) Food and Food Ingredients: Enumeration of Indicator/Spoilage Microorganisms, Media Testing
Packaging Type Plastic Canister
Quantity 10 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.