Learn More
ATCC Aspergillus brasiliensis strain WLRI 034(120) (ATCC 16404™)
Catalog No. 502382512
Encompass
Aspergillus brasiliensis strain WLRI 034(120) (ATCC 16404™) is a whole-genome sequenced fungal strain used in antimicrobial, disinfectant, and preservative efficacy testing. This strain is recommended as a test organism in USP <51>, <61>, <71>, and <2021>.
Recommended companion products:
MicroQuant™ Aspergillus brasiliensis, low CFU (Fisher Cat. No. NC3456088, ATCC 16404-LQ-PACK™),
MicroQuant™ Aspergillus brasiliensis, high CFU (Fisher Cat. No. NC3456077, ATCC 16404-HQ-PACK™)
Genomic DNA (Fisher Cat. No. 50-238-4967, ATCC 16404D-2™).
Catalog No. 50-238-2512 Supplier ATCC Supplier No. 16404
Aspergillus brasiliensis strain WLRI 034(120) (ATCC 16404™) is a whole-genome sequenced fungal strain used in antimicrobial, disinfectant, and preservative efficacy testing. This strain is recommended as a test organism in USP <51>, <61>, <71>, and <2021>.
Recommended companion products:
MicroQuant™ Aspergillus brasiliensis, low CFU (Fisher Cat. No. NC3456088, ATCC 16404-LQ-PACK™),
MicroQuant™ Aspergillus brasiliensis, high CFU (Fisher Cat. No. NC3456077, ATCC 16404-HQ-PACK™)
Genomic DNA (Fisher Cat. No. 50-238-4967, ATCC 16404D-2™).