Learn More
ATCC Acinetobacter radioresistens
Catalog No. 502380416
Encompass
Acinetobacter radioresistens strain IAM 13186 [DSM 6976, FO-1] is a bacterial type strain that was isolated in 1978 from a cotton tampon. The depositor states that this strain is radiation resistant.
Catalog No. 50-238-0416 Supplier ATCC Supplier No. 43998
 Restricted Item
Add to Cart
Add to Cart
Acinetobacter radioresistens strain IAM 13186 [DSM 6976, FO-1] is a bacterial type strain that was isolated in 1978 from a cotton tampon. The depositor states that this strain is radiation resistant.