Learn More
ATCC Quantitative Genomic DNA from; Aspergillus terreus; Strain: MF4845
Catalog No. 502380955
Encompass
Quantitative Genomic DNA from Aspergillus terreus strain MF4845 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-238-0955 Supplier ATCC Supplier No. 20542DQ
Quantitative Genomic DNA from Aspergillus terreus strain MF4845 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.