Learn More
ATCC Aspergillus niger
Supplier:  ATCC 10577
Encompass
Aspergillus niger strain IMI 27809 is provided as an ampoule containing viable cells (may include spores and mycelia) suspended in cryoprotectant. This strain is cited to produce citric acid from brewery wastes, figs, and beet molasses.
Catalog No. 50-238-2516
 Restricted Item