Learn More
ATCC Aspergillus versicolor; MIT-1c; genomic DNA
Supplier:  ATCC 11730D2
Encompass
Genomic DNA isolated from Aspergillus versicolor QM 432. This fungal strain is also available as ATCC Catalog No.: 11730™.
Catalog No. 50-238-2262