Learn More
Cayman Chemical Hirsutide, Purity: ≥95%, CAS: 865368-30-5, 5 mg
SDP
Catalog No. 502934409
Encompass_Preferred
A cyclotetrapeptide fungal metabolite; active against C. pyogenes, S. aureus, P. aeruginosa, and K. pneumonia bacteria (MICs = 25, 13, 6, and 21 μg/cm3, respectively); active against C. albicans, M. audouinii, A. niger, and Ganoderma fungi (MICs = 13, 6, 25, and 6 μg/cm3, respectively)
Catalog No. 50-293-4409 Supplier Cayman Chemical Supplier No. 250145 MG
A cyclotetrapeptide fungal metabolite; active against C. pyogenes, S. aureus, P. aeruginosa, and K. pneumonia bacteria (MICs = 25, 13, 6, and 21 μg/cm3, respectively); active against C. albicans, M. audouinii, A. niger, and Ganoderma fungi (MICs = 13, 6, 25, and 6 μg/cm3, respectively)