Learn More
AdipoGen Asperlactone
SDP
Supplier:  AdipoGen BVT0260M001
Encompass_Preferred
Chemical. CAS 76375-62-7. Formula C9H12O4. MW 184.2. Isolated from Aspergillus ochraceus. Antibiotic. Nematicidal, insecticidal, antibacterial and antifungal.
Catalog No. 50-256-2031