Learn More
Perkin Elmer US LLC Asperon Spray Chamber
Catalog No. 502112922
Encompass_Preferred
The NexION ICP-MS Single Cell Analysis System, with its Asperon Single Cell Spray Chamber, Single Cell Micro DX Autosampler, and Syngistix Single Cell Software Module, offers a first-to-market complete solution to quantitatively measure the metal content in individual cells. The Asperon Spray Chamber is part of our Single Cell Sample Introduction Kits: Single Cell Kit for NexION 1000/2000 (N8150032) and Single Cell Kit for NexION 300/350 (N8140032).
Catalog No. 50-211-2922 Supplier Perkin Elmer US LLC Supplier No. N8152493
The NexION ICP-MS Single Cell Analysis System, with its Asperon Single Cell Spray Chamber, Single Cell Micro DX Autosampler, and Syngistix Single Cell Software Module, offers a first-to-market complete solution to quantitatively measure the metal content in individual cells. The Asperon Spray Chamber is part of our Single Cell Sample Introduction Kits: Single Cell Kit for NexION 1000/2000 (N8150032) and Single Cell Kit for NexION 300/350 (N8140032).