Learn More
Cayman Chemical AsperosaponIn VI 50mg
SDP
Supplier:  Cayman Chemical 3320650 MG
Encompass_Preferred
A triterpenoid saponin; increases BMP2 levels in, and induces differentiation of, MC3T3-E1 preosteoblasts in a concentration-dependent manner; reduces joint inflammation and bone loss in a mouse model of collagen-induced arthritis at 20 mg/kg; reduces increases in acetic acid-induced vascular permeability and increases the latency to paw withdrawal in the hot plate test in mice at 150 and 450 mg/kg
Catalog No. 50-294-7232