Learn More
Medchemexpress LLC HY-N0265 5mg Medchemexpress, Asperosaponin VI CAS:39524-08-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02655MG
Encompass_Preferred
Medchemexpress, HY-N0265 5mg Asperosaponin VI CAS:39524-08-8 Asperosaponin VI, A saponin component from Dipsacus asper wall, induces osteoblast differentiation through BMP‐2/p38 and ERK1/2 pathway. Asperosaponin Ⅵ inhibits apoptosis in hypoxia-induced cardiomyocyte by increasing the Bcl-2/Bax ratio and decreasing active caspase-3 expression, as well as enhancing of p-Akt and p-CREB. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.