Learn More
AdipoGen Asperphenamate (1 mg)
SDP
Supplier:  AdipoGen AGCN20171M001
Encompass_Preferred
Chemical. CAS: 63631-36-7. Formula: C32H30N2O4. MW: 506.6. Isolated from Aspergillus sp. Anticancer compound. Cytotoxic against human breast cancer cells. Induces autophagic cell death in MCF-7 cells. Moderate radical scavenger. Weak acetylcholinesterase (AChE) inhibitor. Shows moderate trypanocidal activity.
Catalog No. 50-205-3564