Learn More
Cayman Chemical Asperphenamate, Purity: ≥95%, CAS: 63631-36-7, 5 mg
SDP
Catalog No. 502935263
Encompass_Preferred
A fungal secondary metabolite; inhibits proliferation of T47D and MDA-MB-231 breast and HL-60 leukemia cancer cell lines (IC50s = 92.3, 96.5, and 97.9 μM, respectively); inhibits yeast α-glucosidase activity (IC50 = 8.3 μM)
Catalog No. 50-293-5263 Supplier Cayman Chemical Supplier No. 250865 MG
A fungal secondary metabolite; inhibits proliferation of T47D and MDA-MB-231 breast and HL-60 leukemia cancer cell lines (IC50s = 92.3, 96.5, and 97.9 μM, respectively); inhibits yeast α-glucosidase activity (IC50 = 8.3 μM)