Learn More
Cayman Chemical Asperuloside, Purity: ≥98%, CAS: 14259-45-1, 10 mg
SDP
Catalog No. 502991617
Encompass_Preferred
An iridoid glycoside with diverse biological activities; reduces microvessel formation by 67% in a chick embryo chorioallantoic membrane assay at 2 μg/egg; inhibits LPS-induced increases in TNF-α, IL-1β, and IL-6 production in RAW 264.7 macrophages in a concentration-dependent manner at 20, 40, and 80 mg/L; reduces lung MPO activity and BALF levels of TNF-α, IL-1β, and IL-6 in a mouse model of LPS-induced acute lung injury at 20, 40, and 80 mg/kg; dietary administration decreases body weight gain, WAT weight, and the ratio of WAT weight to body weight in a high-fat diet-induced mouse model of metabolic syndrome
Catalog No. 50-299-1617 Supplier Cayman Chemical Supplier No. 2943910 MG
An iridoid glycoside with diverse biological activities; reduces microvessel formation by 67% in a chick embryo chorioallantoic membrane assay at 2 μg/egg; inhibits LPS-induced increases in TNF-α, IL-1β, and IL-6 production in RAW 264.7 macrophages in a concentration-dependent manner at 20, 40, and 80 mg/L; reduces lung MPO activity and BALF levels of TNF-α, IL-1β, and IL-6 in a mouse model of LPS-induced acute lung injury at 20, 40, and 80 mg/kg; dietary administration decreases body weight gain, WAT weight, and the ratio of WAT weight to body weight in a high-fat diet-induced mouse model of metabolic syndrome