Learn More
Medchemexpress LLC Asperulosidic Acid | 25368-11-0 | 25 MG
SDP
Catalog No. 5000420380
Encompass_Preferred
Asperulosidic Acid (ASPA) is a bioactive iridoid glycoside extracted from the herbs of *Hedyotis diffusa Willd*. It possesses anti-tumor, anti-oxidant, and anti-inflammatory activities. ASPA's anti-inflammatory effects are linked to the inhibition of inflammatory cytokines (TNF-α, IL-6) and mediators by suppressing the NF-κB and mitogen-activated protein kinase (MAPK) signaling pathways.
  • Bioactive iridoid glycoside
  • Extracted from the herbs of Hedyotis diffusa Willd
  • Exhibits anti-tumor activity
  • Exhibits anti-oxidant activity
  • Exhibits anti-inflammatory activity
  • Inhibits inflammatory cytokines (TNF-α, IL-6)
  • Suppresses NF-κB and MAPK signaling pathways
  • Purity of 99.63%
  • White to off-white solid
Catalog No. 50-004-20380 Supplier Medchemexpress LLC Supplier No. 25MGHYN6246
Add to Cart
Add to Cart
Asperulosidic Acid (ASPA) is a bioactive iridoid glycoside extracted from the herbs of *Hedyotis diffusa Willd*. It possesses anti-tumor, anti-oxidant, and anti-inflammatory activities. ASPA's anti-inflammatory effects are linked to the inhibition of inflammatory cytokines (TNF-α, IL-6) and mediators by suppressing the NF-κB and mitogen-activated protein kinase (MAPK) signaling pathways.
  • Bioactive iridoid glycoside
  • Extracted from the herbs of Hedyotis diffusa Willd
  • Exhibits anti-tumor activity
  • Exhibits anti-oxidant activity
  • Exhibits anti-inflammatory activity
  • Inhibits inflammatory cytokines (TNF-α, IL-6)
  • Suppresses NF-κB and MAPK signaling pathways
  • Purity of 99.63%
  • White to off-white solid