Learn More
Medchemexpress LLC Asperulosidic acid | 25368-11-0 | MFCD21333373 | 1 MG
SDP
Catalog No. 5000418831
Encompass_Preferred
Asperulosidic acid (ASPA) is a bioactive iridoid glycoside extracted from the herbs of Hedyotis diffusa Willd. It exhibits anti-tumor, anti-oxidant, and anti-inflammatory activities. ASPA's activities are linked to the inhibition of inflammatory cytokines (TNF-α, IL-6) and mediators through the suppression of the NF-κB and mitogen-activated protein kinase (MAPK) signaling pathways.
  • Bioactive iridoid glycoside
  • Extracted from *Hedyotis diffusa Willd*
  • Anti-tumor activity
  • Anti-oxidant activity
  • Anti-inflammatory activity
  • Suppresses NF-κB and MAPK signaling pathways
  • Down-regulates mRNA levels of TNF-α and IL-6 in LPS-induced RAW 264.7 cells
  • Decreases IκB-α phosphorylation
  • Reduces Erk1/2 phosphorylation
Catalog No. 50-004-18831 Supplier Medchemexpress LLC Supplier No. HYN62461MG
Asperulosidic acid (ASPA) is a bioactive iridoid glycoside extracted from the herbs of Hedyotis diffusa Willd. It exhibits anti-tumor, anti-oxidant, and anti-inflammatory activities. ASPA's activities are linked to the inhibition of inflammatory cytokines (TNF-α, IL-6) and mediators through the suppression of the NF-κB and mitogen-activated protein kinase (MAPK) signaling pathways.
  • Bioactive iridoid glycoside
  • Extracted from *Hedyotis diffusa Willd*
  • Anti-tumor activity
  • Anti-oxidant activity
  • Anti-inflammatory activity
  • Suppresses NF-κB and MAPK signaling pathways
  • Down-regulates mRNA levels of TNF-α and IL-6 in LPS-induced RAW 264.7 cells
  • Decreases IκB-α phosphorylation
  • Reduces Erk1/2 phosphorylation