Learn More
Aviva Systems Biology Corp ASPH Antibody - N-terminal region (ARP51357_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51357_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ASPH. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ASPH. Alternate names: BAH, CASQ2BP1, HAAH, JCTN, junctin, AAHApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60559