Learn More
Aviva Systems Biology Corp ASPH Antibody - middle region (ARP34691_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34691_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASPH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ASPH. Alternate names: BAH, CASQ2BP1, HAAH, JCTN, junctin, AAHApplicationWB, IHC. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-55514