Learn More
Alkali Scientific ASPIR-8™, 25mL Reservoir for 8-channel Pipettes, Bulk Pack, 100/cs
SDP
Catalog No. 502389231
Encompass_Preferred
ASPIR-8™ is the ideal reservoir for any lab utilizing 8-channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over their longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPIR-8 is the only reservoir basin that is tailor-made for your 8-channel instrument. 100 per pack.
Catalog No. 50-238-9231 Supplier Alkali Scientific Supplier No. RS88100
ASPIR-8™ is the ideal reservoir for any lab utilizing 8-channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over their longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPIR-8 is the only reservoir basin that is tailor-made for your 8-channel instrument. 100 per pack.