Learn More
Thomas Scientific Aspir-8 Reservoir, 25ml, 8-channel only, sterile, 5/sterile bag
SDP
Catalog No. NC1603507
Encompass
Our new ASPIR-8™ is the ideal reservoir for any lab utilizing 8 channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over its longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPI-8 is the only reservoir that is tailor-made for your 8-channel instrument.
Catalog No. NC1603507 Supplier Thomas Scientific Supplier No. 1153Z92
Our new ASPIR-8™ is the ideal reservoir for any lab utilizing 8 channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over its longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPI-8 is the only reservoir that is tailor-made for your 8-channel instrument.