Learn More
Med Vet International Aspir-Guard, Set, 3 Pack of all 3 Sizes, J1494S, 3/Pack
Supplier:  Med Vet International JOR1494S
Encompass_Preferred
Prevent accidental aspiration Aspir-Guard prevents fluids and dental debris from being accidentally aspirated during a routine dental procedure. Aspir-Guard is a dehydrated PVA donut shaped product. Attached strings are used to tie the Aspir-Guard to the roll gauze or old IV tubing used to secure the ET tube to the patient. After the patient is intubated, the Aspir-Guard is pushed into place to block the oropharynx from fluid aspiration. Once the procedure is finished, the Aspir-Guard is removed gently together with the ET tube. Three of each size to a package. Non-sterile. PVA (Polyvinyl Alcohol) Set of all 3 sizes includes: J1494a - small 20mm 30m Hole size: 6mm, fits tubes 3.5-6.0mm ID J1494b - medium 25mm 40mm Hole size: 9mm, fits tubes 6.5-8.0mm ID J1494c - large 30mm 50mm Hole size: 12mm, fits tubes 9.0-10.0mm ID
Catalog No. 50-005-85820