Learn More
Bellco Glass, Inc. Aspirating Bulb
Supplier: Bellco Glass, Inc. 229279000
DescriptionAspirating bulb for use with Aspirating and Plasteur Pipets Pack Description12/BagCase Quantity12SterileNoAutoclavableYesNon-pyrogenicYesPrimary ColorBlackPrimary MaterialRubberVolume1mLRNase/DNase FreeYes