Learn More
Quality Biological Inc Aspirating Pipet, 1 mL, Sterile, bulk packed (not individually wrapped), 50 per bag, 400/CS
SDP
Catalog No. 507517910
Encompass_Preferred
Aspirating Pipet, 1 mL, Sterile, Bulk Packed (not individually wrapped)
Packaged 25 Bulk Packed/Bag, 400/CS
• Ideal for aspiration of media and sample waste removal without needing to measure the volume
• Supplied without graduations or plugs
Catalog No. 50-751-7910 Supplier Quality Biological Inc Supplier No. P886229251
Aspirating Pipet, 1 mL, Sterile, Bulk Packed (not individually wrapped)
Packaged 25 Bulk Packed/Bag, 400/CS
• Ideal for aspiration of media and sample waste removal without needing to measure the volume
• Supplied without graduations or plugs